1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
1-tert-Butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate, Ethyl 1-Boc-3-oxopiperidine-4-carboxylate
069342-5G
069342-25G
069342-1G
101939-712EA
50.77
USD
101939-712
102551-744
102833-462
1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate ≥95%
1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate
Hazard: Irritant/Refrigerate
Formula:
C₁₃H₂₁NO₅ MW: 271.31 g/mol |
MDL Number:
MFCD09878815 CAS Number: 71233-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...