1-(1-Benzylpiperidin-3-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: Matrix Scientific

069339-250MG 069339-500MG
101939-702EA 727.69 USD
101939-702 101939-704
1-(1-Benzylpiperidin-3-yl)-2,2,2-trifluoroethanone ≥95%
1-(1-Benzylpiperidin-3-yl)-2,2,2-trifluoroethanone

Hazard: Irritant

Formula: C₁₄H₁₆F₃NO
MDL Number: MFCD18909669
CAS Number: 1182349-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR