Methyl-1-benzylpiperidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

069338-1G 069338-5G
101939-698EA 698.46 USD
101939-698 101939-700
Methyl-1-benzylpiperidine-3-carboxylate ≥95%
Methyl-1-benzylpiperidine-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₉NO₂
MDL Number: MFCD15146668
CAS Number: 50585-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR