3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride ≥96%

Supplier: Matrix Scientific

Synonyms: (3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride, 3-aminomethylphenylboronic acid, pinacol ester hydrochloride

069336-1G 069336-5G
101939-690EA 698.46 USD
101939-690 101939-692
3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride ≥96%
3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁BClNO₂
MW: 269.58 g/mol
MDL Number: MFCD03410294
CAS Number: 380430-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR