Potassium-5-(methoxycarbonyl)nicotinate ≥95%

Supplier: Matrix Scientific

069335-1G 069335-5G
101939-686EA 698.46 USD
101939-686 101939-688
Potassium-5-(methoxycarbonyl)nicotinate ≥95%
Potassium-5-(methoxycarbonyl)nicotinate

Hazard: Irritant

Formula: C₈H₆KNO₄
MDL Number: MFCD12026433
CAS Number: 27247-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR