1-Trifluoromethyl-1,2-benziodoxol-3(1H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: Togni Reagent II

069332-250MG 069332-1G
101939-672EA 353.08 USD
101939-672 101939-676
1-Trifluoromethyl-1,2-benziodoxol-3(1H)-one ≥95%
1-Trifluoromethyl-1,2-benziodoxol-3(1H)-one

Hazard: Irritant

Formula: C₈H₄F₃IO₂
MW: 316.02 g/mol
MDL Number: MFCD18800706
CAS Number: 887144-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR