1-Benzylpyrrolidine-3-carbonitrile oxalate ≥95%

Supplier: Matrix Scientific

069325-1G 069325-5G
101939-652EA 378.46 USD
101939-652 101939-654
1-Benzylpyrrolidine-3-carbonitrile oxalate ≥95%
1-Benzylpyrrolidine-3-carbonitrile oxalate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₄
MDL Number: MFCD11501670
CAS Number: 1188265-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR