tert-Butyl-4-(1H-indol-3-yl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(1H-indol-3-yl)piperidine-1-carboxylate

069322-250MG 069322-1G
101939-648EA 698.46 USD
101939-648 101939-650
tert-Butyl-4-(1H-indol-3-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(1H-indol-3-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₂
MW: 300.4 g/mol
MDL Number: MFCD11840339
CAS Number: 155302-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR