3-(Aminomethyl)phenylboronic acid hydrochloride ≥96%
Supplier: Matrix Scientific
Synonyms:
(3-(Aminomethyl)phenyl)boronic acid hydrochloride, 3-(Aminomethyl)benzeneboronic acid hydrochloride
069319-1G
069319-5G
101939-640EA
395.38
USD
101939-640
101939-642
3-(Aminomethyl)phenylboronic acid hydrochloride ≥96%
3-(Aminomethyl)phenylboronic acid hydrochloride
Hazard: Irritant
Formula:
C₇H₁₁BClNO₂ MW: 187.43 g/mol |
MDL Number:
MFCD02093050 CAS Number: 352525-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...