3-(Hydroxymethyl)-1-adamantanol ≥95%

Synonyms: 3-(Hydroxymethyl)adamantan-1-ol, 1-Hydroxy-3-(hydroxymethyl)adamantane

069317-1G 069317-5G
101939-630EA 2181.54 USD
101939-630 101939-632
3-(Hydroxymethyl)-1-adamantanol ≥95%
3-(Hydroxymethyl)-1-adamantanol

Hazard: Irritant

Formula: C₁₁H₁₈O₂
MW: 182.26 g/mol
MDL Number: MFCD03105096
CAS Number: 38584-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR