4-Methoxytricyclo[3.3.1.13,7]dec-3-ene-1-carboxylic acid methyl ester ≥95%
Supplier: Matrix Scientific
069316-250MG
069316-500MG
101939-626EA
698.46
USD
101939-626
101939-628
4-Methoxytricyclo[3.3.1.13,7]dec-3-ene-1-carboxylic acid methyl ester ≥95%
4-Methoxytricyclo[3.3.1.13,7]dec-3-ene-1-carboxylic acid methyl ester
Hazard: Irritant
Formula:
C₁₂H₁₆O₃ |
MDL Number:
MFCD24849238 CAS Number: 1283717-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...