4,4-Difluoroadamantane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

069313-250MG 069313-500MG
101939-614EA 1309.23 USD
101939-614 101939-616
4,4-Difluoroadamantane-1-carboxylic acid ≥95%
4,4-Difluoroadamantane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₄F₂O₂
MDL Number: MFCD02712630
CAS Number: 438017-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR