Ethyl-1-benzyl-2,4-dioxopiperidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

069310-1G 069310-5G
101939-610EA 615.38 USD
101939-610 101939-612
Ethyl-1-benzyl-2,4-dioxopiperidine-3-carboxylate ≥95%
Ethyl-1-benzyl-2,4-dioxopiperidine-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₇NO₄
MDL Number: MFCD00030997
CAS Number: 198417-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR