1-Benzyl-4-(3-phenylpropyl)-1,2,3,6-tetrahydropyridine ≥95%

Supplier: Matrix Scientific

069309-250MG 069309-500MG
101939-606EA 436.92 USD
101939-606 101939-608
1-Benzyl-4-(3-phenylpropyl)-1,2,3,6-tetrahydropyridine ≥95%
1-Benzyl-4-(3-phenylpropyl)-1,2,3,6-tetrahydropyridine

Hazard: Irritant

Formula: C₂₁H₂₅N
MDL Number: MFCD18800726
CAS Number: 70152-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR