1'-Benzyl-5-fluorospiro[indoline-3,4'-piperidine]-2-one ≥95%

Supplier: Matrix Scientific

069306-250MG 069306-500MG
101939-598EA 989.23 USD
101939-598 101939-600
1'-Benzyl-5-fluorospiro[indoline-3,4'-piperidine]-2-one ≥95%
1'-Benzyl-5-fluorospiro[indoline-3,4'-piperidine]-2-one

Hazard: Irritant

Formula: C₁₉H₁₉FN₂O
MDL Number: MFCD18433704
CAS Number: 1258638-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR