5-(Methoxycarbonyl)piperidine-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
069303-250MG
069303-500MG
101939-590EA
669.23
USD
101939-590
101939-592
5-(Methoxycarbonyl)piperidine-3-carboxylic acid ≥95%
5-(Methoxycarbonyl)piperidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₈H₁₃NO₄ |
MDL Number:
MFCD12026400 CAS Number: 748113-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...