Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-benzyl-5,5-difluoro-4-oxopiperidine-3-carboxylate

069302-250MG 069302-500MG 069302-1G
101939-584EA 930.77 USD
101939-584 101939-586 101939-588
Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate ≥95%
Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₇F₂NO₃
MW: 297.3 g/mol
MDL Number: MFCD16036533
CAS Number: 1067915-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR