tert-Butyl-4-(6-formylpyrazolo[1,5-a]pyrimidin-2-yl)piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
069296-250MG
069296-500MG
101939-580EA
669.23
USD
101939-580
101939-582
tert-Butyl-4-(6-formylpyrazolo[1,5-a]pyrimidin-2-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(6-formylpyrazolo[1,5-a]pyrimidin-2-yl)piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₇H₂₂N₄O₃ MW: 330.39 g/mol |
MDL Number:
MFCD18433797 CAS Number: 1258638-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...