Ethyl-5-ethyl-4-hydroxypyrrolo[1,2-f][1,2,4]triazine-6-carboxylate ≥95%

Supplier: Matrix Scientific

069291-500MG 069291-1G 069291-5G
101939-574EA 769.23 USD
101939-574 101939-576 101939-578
Ethyl-5-ethyl-4-hydroxypyrrolo[1,2-f][1,2,4]triazine-6-carboxylate ≥95%
Ethyl-5-ethyl-4-hydroxypyrrolo[1,2-f][1,2,4]triazine-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₃
MW: 235.24 g/mol
MDL Number: MFCD12756029
CAS Number: 310436-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR