(3R,4R)-4-Amino-1-[[4-[(3-methoxyphenyl)amino]pyrrolo-[2,1-f][1,2,4]triazin-5-yl]methyl]piperidin-3-ol ≥95%
Supplier: Matrix Scientific
069274-100MG
069274-250MG
069274-500MG
101939-560EA
2850.77
USD
101939-560
101939-562
101939-564
(3R,4R)-4-Amino-1-[[4-[(3-methoxyphenyl)amino]pyrrolo-[2,1-f][1,2,4]triazin-5-yl]methyl]piperidin-3-ol ≥95%
(3R,4R)-4-Amino-1-[[4-[(3-methoxyphenyl)amino]pyrrolo-[2,1-f][1,2,4]triazin-5-yl]methyl]piperidin-3-ol
Hazard: Irritant
Formula:
C₁₉H₂₄N₆O₂ |
MDL Number:
MFCD18633295 CAS Number: 859853-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...