4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine

069273-250MG 069273-1G
101939-554EA 306.15 USD
101939-554 101939-558
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine ≥95%
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine

Hazard: Irritant

Formula: C₆H₅BrN₄
MW: 213.04 g/mol
MDL Number: MFCD09033848
CAS Number: 937046-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR