2-(4-Methoxyphenyl)tetrahydrofuran ≥95%

Supplier: Matrix Scientific

069271-1G 069271-5G
101939-544EA 1483.08 USD
101939-544 101939-546
2-(4-Methoxyphenyl)tetrahydrofuran ≥95%
2-(4-Methoxyphenyl)tetrahydrofuran

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD00970923
CAS Number: 79623-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR