3-(Bromomethyl)-3-(chloromethyl)oxetane ≥95%

Supplier: Matrix Scientific

069264-250MG 069264-500MG
101939-518EA 989.23 USD
101939-518 101939-520
3-(Bromomethyl)-3-(chloromethyl)oxetane ≥95%
3-(Bromomethyl)-3-(chloromethyl)oxetane

Hazard: Irritant

Formula: C₅H₈BrClO
MDL Number: MFCD18909286
CAS Number: 35842-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR