(3-Phenyloxetan-3-yl)methanol ≥95%

Supplier: Matrix Scientific

069263-250MG 069263-500MG
101939-514EA 872.31 USD
101939-514 101939-516
(3-Phenyloxetan-3-yl)methanol ≥95%
(3-Phenyloxetan-3-yl)methanol

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD18642402
CAS Number: 114012-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR