(3-Phenyloxetan-3-yl)methanamine ≥95%

Supplier: Matrix Scientific

069261-250MG 069261-500MG
101939-506EA 607.69 USD
101939-506 101939-508
(3-Phenyloxetan-3-yl)methanamine ≥95%
(3-Phenyloxetan-3-yl)methanamine

Hazard: Irritant

Formula: C₁₀H₁₃NO
MDL Number: MFCD18251065
CAS Number: 497239-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR