tert-Butyl-5-bromo-1H-indazole-1-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1H-indazole, N1-BOC protected, tert-Butyl 5-bromo-1H-indazole-1-carboxylate

069258-1G 069258-5G
101939-498EA 524.62 USD
101939-498 101939-500
tert-Butyl-5-bromo-1H-indazole-1-carboxylate ≥98%
tert-Butyl-5-bromo-1H-indazole-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃BrN₂O₂
MW: 297.16 g/mol
MDL Number: MFCD08435913
CAS Number: 651780-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR