1-Methylpiperazin-2-one ≥95%

Supplier: Matrix Scientific

069257-1G 069257-5G
101939-494EA 195.38 USD
101939-494 101939-496
1-Methylpiperazin-2-one ≥95%
1-Methylpiperazin-2-one

Hazard: Irritant

Formula: C₅H₁₀N₂O
MW: 114.1457 g/mol
MDL Number: MFCD00102572
CAS Number: 59702-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR