tert-Butyl-3-amino-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected, tert-Butyl 3-amino-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate

069254-1G 069254-5G
101939-490EA 184.62 USD
101939-490 101939-492
tert-Butyl-3-amino-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate ≥95%
tert-Butyl-3-amino-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate

Hazard: Irritant-Harmful/Refrigerate

Formula: C₁₀H₁₆N₄O₂
MW: 224.2596 g/mol
MDL Number: MFCD08275050
CAS Number: 398491-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR