4-Amino-2,6-dimethylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2,6-Dimethylpyridin-4-amine

069253-1G 069253-5G
101939-486EA 60 USD
101939-486 101939-488
4-Amino-2,6-dimethylpyridine ≥98%
4-Amino-2,6-dimethylpyridine

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00130078
CAS Number: 3512-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR