2-Chloropyridine-3,4-diamine ≥98%

Supplier: Matrix Scientific

Synonyms: 3,4-Diamino-2-chloropyridine, 2-Chloropyridine-3,4-diamine

069252-1G 069252-5G
101939-482EA 101.54 USD
101939-482 101939-484
2-Chloropyridine-3,4-diamine ≥98%
2-Chloropyridine-3,4-diamine

Hazard: Irritant

Formula: C₅H₆ClN₃
MW: 143.5742 g/mol
MDL Number: MFCD07363739
CAS Number: 39217-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR