4-(Bromomethyl)tetrahydropyran ≥97%
Supplier: Matrix Scientific
069250-1G
069250-5G
069250-25G
101939-472EA
36.92
USD
101939-472
101939-474
101939-476
4-(Bromomethyl)tetrahydropyran ≥97%
4-(Bromomethyl)tetrahydropyran
Hazard: Corrosive
Formula:
C₆H₁₁BrO MW: 179.0549 g/mol Boiling Pt: 89…93 °C (10 mmHg) |
MDL Number:
MFCD03788561 CAS Number: 125552-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...