5-Isoquinolinecarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Isoquinoline-5-carboxylic acid

069249-1G 069249-5G
101939-468EA 250.77 USD
101939-468 101939-470
5-Isoquinolinecarboxylic acid ≥97%
5-Isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
MDL Number: MFCD03788744
CAS Number: 27810-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR