Isoquinoline-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

069248-1G 069248-5G 069248-25G
101939-462EA 524.62 USD
101939-462 101939-464 101939-466
Isoquinoline-3-carboxylic acid ≥97%
Isoquinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₂
MW: 173.1681 g/mol
MDL Number: MFCD00075137
CAS Number: 6624-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR