Quinoxaline-6-acetic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Quinoxalin-6-yl)acetic acid

069244-1G 069244-5G 069244-25G
101939-444EA 930.77 USD
101939-444 101939-446 101939-448
Quinoxaline-6-acetic acid ≥97%
Quinoxaline-6-acetic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD09880609
CAS Number: 473932-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR