6-Chloro-3-nitroimidazo[1,2-b]pyridazine ≥98%

Supplier: Matrix Scientific

069243-1G 069243-5G
101939-440EA 36.92 USD
101939-440 101939-442
6-Chloro-3-nitroimidazo[1,2-b]pyridazine ≥98%
6-Chloro-3-nitroimidazo[1,2-b]pyridazine

Hazard: Irritant

Formula: C₆H₃ClN₄O₂
MDL Number: MFCD08276991
CAS Number: 18087-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR