Cyclopamine ≥99%
Supplier: Matrix Scientific
Synonyms:
(2'R,3S,3'R,3a'S,6aS,6bS,6'S,7a'R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3a',4,4',5',6,6a,6b,6',7,7',7a',8,11,11a,11b-octadecahydro-3'H-spiro[benzo[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol
069242-25MG
069242-50MG
069242-100MG
101939-434EA
276.92
USD
101939-434
101939-436
101939-438
Cyclopamine ≥99%
Cyclopamine
Hazard: Irritant
Formula:
C₂₇H₄₁NO₂ MW: 411.63 g/mol |
MDL Number:
MFCD09878269 CAS Number: 4449-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...