Ethyl-8-methylimidazo[1,2-a]pyridine-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 8-methylimidazo[1,2-a]pyridine-2-carboxylate

069241-1G 069241-5G
101939-430EA 116.92 USD
101939-430 101939-432
Ethyl-8-methylimidazo[1,2-a]pyridine-2-carboxylate ≥97%
Ethyl-8-methylimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MW: 204.2300 g/mol
MDL Number: MFCD03419470
CAS Number: 67625-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR