Ethyl-6-methylimidazo[1,2-a]pyridine-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-methylimidazo[1,2-a]pyridine-2-carboxylate

069240-1G 069240-5G
101939-426EA 524.62 USD
101939-426 101939-428
Ethyl-6-methylimidazo[1,2-a]pyridine-2-carboxylate ≥97%
Ethyl 6-methylimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MW: 204.2300 g/mol
MDL Number: MFCD02325396
CAS Number: 70705-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR