trans-2-(4-Fluorophenyl)vinylboronic acid ≥98%

Supplier: Matrix Scientific

069237-1G 069237-5G 069237-25G
101939-412EA 175.38 USD
101939-412 101939-414 101939-416
trans-2-(4-Fluorophenyl)vinylboronic acid ≥98%
trans-2-(4-Fluorophenyl)vinylboronic acid

Hazard: Irritant

Formula: C₈H₈BFO₂
MW: 165.96 g/mol
MDL Number: MFCD02093725
CAS Number: 214907-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR