4-Amino-3-bromo-1-methyl-1H-indazole ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-1-methyl-1H-indazol-4-amine

069236-1G 069236-5G
101939-408EA 2384.62 USD
101939-408 101939-410
4-Amino-3-bromo-1-methyl-1H-indazole ≥97%
4-Amino-3-bromo-1-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₈BrN₃
MW: 226.0700 g/mol
MDL Number: MFCD11109355
CAS Number: 1092351-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR