5-Bromo-2-chloromethylimidazo[1,2-a]pyridine ≥96%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-(chloromethyl)imidazo[1,2-a]pyridine

069231-1G 069231-5G
101939-388EA 1803.08 USD
101939-388 101939-390
5-Bromo-2-chloromethylimidazo[1,2-a]pyridine ≥96%
5-Bromo-2-chloromethylimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₆BrClN₂
MW: 245.51 g/mol
MDL Number: MFCD06739242
CAS Number: 885275-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR