Methyl-2-amino-4-oxazolecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-aminooxazole-4-carboxylate

069229-1G 069229-5G
101939-380EA 121.54 USD
101939-380 101939-382
Methyl-2-amino-4-oxazolecarboxylate ≥95%
Methyl 2-amino-4-oxazolecarboxylate

Hazard: Irritant

Formula: C₅H₆N₂O₃
MW: 142.1127 g/mol
MDL Number: MFCD09870069
CAS Number: 1000576-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR