4-Chloro-5-fluoropyrimidine ≥94%
Supplier: Matrix Scientific
069228-1G
069228-5G
101939-376EA
56.92
USD
101939-376
101939-378
4-Chloro-5-fluoropyrimidine ≥94%
4-Chloro-5-fluoropyrimidine
Hazard: Irritant/Refrigerate
Formula:
C₄H₂ClFN₂ |
MDL Number:
MFCD09750157 CAS Number: 347418-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...