tert-Butyl-N-methylcarbamate ≥96%

Supplier: Matrix Scientific

Synonyms: tert-Butyl methylcarbamate

069227-5G 069227-25G
101939-372EA 106.15 USD
101939-372 101939-374
tert-Butyl-N-methylcarbamate ≥96%
tert-Butyl-N-methylcarbamate

Hazard: Irritant/Refrigerate

Formula: C₆H₁₃NO₂
MW: 131.18 g/mol
MDL Number: MFCD08899404
CAS Number: 16066-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR