5-Bromo-2-ethoxypyrimidine ≥99%
Supplier: Matrix Scientific
069223-1G
069223-5G
069223-25G
101939-348EA
175.38
USD
101939-348
101939-350
101939-352
5-Bromo-2-ethoxypyrimidine ≥99%
5-Bromo-2-ethoxypyrimidine
Hazard: Irritant
Formula:
C₆H₇BrN₂O |
MDL Number:
MFCD07375135 CAS Number: 17758-11-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...