4,5-Dimethyl-2-(tributylstannyl)thiazole ≥94%

Supplier: Matrix Scientific

069220-1G 069220-5G
101939-332EA 756.92 USD
101939-332 101939-334
4,5-Dimethyl-2-(tributylstannyl)thiazole ≥94%
4,5-Dimethyl-2-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₇H₃₃NSSn
MDL Number: MFCD09265485
CAS Number: 938181-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR