3-Ethylphenylboronic acid ≥98%

Supplier: Matrix Scientific

069217-1G 069217-5G
101939-318EA 403.08 USD
101939-318 101939-320
3-Ethylphenylboronic acid ≥98%
3-Ethylphenylboronic acid

Hazard: Irritant

Formula: C₈H₁₁BO₂
MDL Number: MFCD06657721
CAS Number: 90555-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR