4-Bromo-2-isopropylthiazole ≥98%

Supplier: Matrix Scientific

069215-500MG 069215-1G 069215-5G
101939-306EA 366.15 USD
101939-306 101939-308 101939-310
4-Bromo-2-isopropylthiazole ≥98%
4-Bromo-2-isopropylthiazole

Hazard: Irritant

Formula: C₆H₈BrNS
MW: 206.11 g/mol
MDL Number: MFCD06660118
CAS Number: 376585-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR