Diethyl (2-chlorothiazol-5-yl)methylphosphonate ≥97%

Supplier: Matrix Scientific

069214-500MG 069214-1G
101939-302EA 1106.15 USD
101939-302 101939-304
Diethyl (2-chlorothiazol-5-yl)methylphosphonate ≥97%
Diethyl (2-chlorothiazol-5-yl)methylphosphonate

Hazard: Irritant

Formula: C₈H₁₃ClNO₃PS
MDL Number: MFCD06660117
CAS Number: 903130-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR