4-Bromoindole-3-carboxyaldehyde ≥97%

Supplier: Matrix Scientific

069212-1G 069212-5G 069212-25G
101939-290EA 116.92 USD
101939-290 101939-292 101939-294
4-Bromoindole-3-carboxyaldehyde ≥97%
4-Bromoindole-3-carboxyaldehyde

Hazard: Irritant

Formula: C₉H₆BrNO
MW: 224.06 g/mol
MDL Number: MFCD05864695
CAS Number: 98600-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR